ADVH49B81EC1 AChemBlock

6-Chloro-4-methoxy-1H-indazole

  • Cas Number : 885519-62-0
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-methoxy-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72404
Availability details for
Quantity
  • Availability
  • Add to Cart