ADVH49B81EC7 AChemBlock

trans-2-(difluoromethyl)cyclopropanecarboxylic acid

  • Cas Number : 883443-58-1
  • Molecular Weight : 136.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-2-difluoromethyl-cyclopropanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73494
Availability details for
Quantity
  • Availability
  • Add to Cart