ADVH49B81EC9 AChemBlock

Methyl2-aMino-5-nitronicotinate

  • Cas Number : 88312-64-5
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl2-amino-5-nitronicotinate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86501
Availability details for
Quantity
  • Availability
  • Add to Cart