ADVH49B81ECF AChemBlock

2-(Methylsulfonyl)phenylboronic acid pinacol ester

  • Cas Number : 879648-22-3
  • Molecular Weight : 300.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/2-methylsulfonyl-phenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70588
Availability details for
Quantity
  • Availability
  • Add to Cart