ADVH49B81ED0 AChemBlock

1-Isopropyl-1H-pyrazol-4-boronic acid, pinacol ester

  • Cas Number : 879487-10-2
  • Molecular Weight : 254.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-isopropyl-1h-pyrazol-4-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70721
Availability details for
Quantity
  • Availability
  • Add to Cart