ADVH49B81ED1 AChemBlock

Eprobemide

  • Cas Number : 87940-60-1
  • Molecular Weight : 282.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/eprobemide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86664
Availability details for
Quantity
  • Availability
  • Add to Cart