ADVH49B81ED3 AChemBlock

O-(4-methoxybenzyl)hydroxylamine hydrochloride

  • Cas Number : 876-33-5
  • Molecular Weight : 189.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/o-4-methoxybenzyl-hydroxylamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70993
Availability details for
Quantity
  • Availability
  • Add to Cart