ADVH49B81ED4 AChemBlock

2-Fluorophenylboronic acid, pinacol ester

  • Cas Number : 876062-39-4
  • Molecular Weight : 240.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70607
Availability details for
Quantity
  • Availability
  • Add to Cart