ADVH49B81ED5 AChemBlock

(4S,5S)-5-[3,5-bis(trifluoromethyl)phenyl]-4-methyl-1,3-oxazolidin-2-one

  • Cas Number : 875444-06-7
  • Molecular Weight : 313.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4s-5s-5-3-5-bis-trifluoromethyl-phenyl-4-methyl-1-3-oxazolidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73592
Availability details for
Quantity
  • Availability
  • Add to Cart