ADVH49B81ED6 AChemBlock

Quisinostat dihydrochloride

  • Cas Number : 875320-31-3
  • Molecular Weight : 467.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/quisinostat-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73593
Availability details for
Quantity
  • Availability
  • Add to Cart