ADVH49B81ED7 AChemBlock

6-fluoro-5-(trifluoromethyl)-1H-indole

  • Cas Number : 875306-79-9
  • Molecular Weight : 203.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-5-trifluoromethyl-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73595
Availability details for
Quantity
  • Availability
  • Add to Cart