ADVH49B81EE2 AChemBlock

4,4,4-trifluoro-3-hydroxybutanoic acid

  • Cas Number : 86884-21-1
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-4-trifluoro-3-hydroxybutanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73657
Availability details for
Quantity
  • Availability
  • Add to Cart