ADVH49B81EE4 AChemBlock

3,9-Diazaspiro[5.5]undecan-2-one

  • Cas Number : 867006-20-0
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-9-diazaspiro-5-5-undecan-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73664
Availability details for
Quantity
  • Availability
  • Add to Cart