ADVH49B81EE5 AChemBlock

5-(4-methoxy-1-piperidyl)pyridin-2-amine

  • Cas Number : 866620-43-1
  • Molecular Weight : 207.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-4-methoxy-1-piperidyl-pyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73668
Availability details for
Quantity
  • Availability
  • Add to Cart