ADVH49B81EEB AChemBlock

4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridazine

  • Cas Number : 863422-41-7
  • Molecular Weight : 206.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-tetramethyl-1-3-2-dioxaborolan-2-yl-pyridazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73704
Availability details for
Quantity
  • Availability
  • Add to Cart