ADVH49B81EEC AChemBlock

5-methoxy-3,4-dihydro-2H-1-benzopyran-4-one

  • Cas Number : 863309-86-8
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-3-4-dihydro-2h-1-benzopyran-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73707
Availability details for
Quantity
  • Availability
  • Add to Cart