ADVH49B81EF1 AChemBlock

1-(5-(benzyloxy)-2-hydroxy-3-nitrophenyl)ethanone

  • Cas Number : 861841-94-3
  • Molecular Weight : 287.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.achemblock.com/1-5-benzyloxy-2-hydroxy-3-nitrophenyl-ethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86461
Availability details for
Quantity
  • Availability
  • Add to Cart