ADVH49B81EF6 AChemBlock

6-aminoquinoline-4-carboxylic acid

  • Cas Number : 859924-25-7
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-aminoquinoline-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73730
Availability details for
Quantity
  • Availability
  • Add to Cart