ADVH49B81EF7 AChemBlock

6-methyl-1H-indazole-3-carboxylic acid

  • Cas Number : 858227-12-0
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-1h-indazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73741
Availability details for
Quantity
  • Availability
  • Add to Cart