ADVH49B81F01 AChemBlock

4-bromo-5,6,7,8-tetrahydroisoquinoline

  • Cas Number : 850349-00-7
  • Molecular Weight : 212.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-5-6-7-8-tetrahydroisoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73821
Availability details for
Quantity
  • Availability
  • Add to Cart