ADVH49B81F06 AChemBlock

2-((1R,4R)-4-((tert-butyldimethylsilyl)oxy)cyclohexyl)isoindoline-1,3-dione

  • Cas Number : 847416-23-3
  • Molecular Weight : 359.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-1r-4r-4-tert-butyldimethylsilyl-oxy-cyclohexyl-isoindoline-1-3-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86710
Availability details for
Quantity
  • Availability
  • Add to Cart