ADVH49B81F08 AChemBlock

6,7-difluoroquinoxaline-2-carboxylic acid

  • Cas Number : 845782-63-0
  • Molecular Weight : 210.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-7-difluoroquinoxaline-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73869
Availability details for
Quantity
  • Availability
  • Add to Cart