ADVH49B81F09 AChemBlock

7-methoxy-2,3-dihydro-1H-indole-2,3-dione

  • Cas Number : 84575-27-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-methoxy-2-3-dihydro-1h-indole-2-3-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73872
Availability details for
Quantity
  • Availability
  • Add to Cart