ADVH49B81F0A AChemBlock

2-methyl-5-nitropyridin-4-amine

  • Cas Number : 84487-12-7
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-5-nitropyridin-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73880
Availability details for
Quantity
  • Availability
  • Add to Cart