ADVH49B81F0C AChemBlock

methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

  • Cas Number : 844500-75-0
  • Molecular Weight : 277.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86350
Availability details for
Quantity
  • Availability
  • Add to Cart