ADVH49B81F0D AChemBlock

2-(3-amino-1H-pyrazol-1-yl)ethan-1-ol

  • Cas Number : 84407-13-6
  • Molecular Weight : 127.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-amino-1h-pyrazol-1-yl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73883
Availability details for
Quantity
  • Availability
  • Add to Cart