ADVH49B81F0E AChemBlock

4-(ethylamino)piperidine-4-carboxamide

  • Cas Number : 84100-54-9
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-ethylamino-piperidine-4-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73897
Availability details for
Quantity
  • Availability
  • Add to Cart