ADVH49B81F0F AChemBlock

(E)-1-propenyl boronic pinacol ester

  • Cas Number : 83947-58-4
  • Molecular Weight : 186.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/e-1-propenyl-boronic-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72328
Availability details for
Quantity
  • Availability
  • Add to Cart