ADVH49B81F12 AChemBlock

tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline-1-carboxylate

  • Cas Number : 837392-67-3
  • Molecular Weight : 345.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-indoline-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73910
Availability details for
Quantity
  • Availability
  • Add to Cart