ADVH49B81F14 AChemBlock

2-Amino-1,3-thiazole-4-carboxylic acid, N-BOC protected

  • Cas Number : 83673-98-7
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-1-3-thiazole-4-carboxylic-acid-n-boc-protected.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71046
Availability details for
Quantity
  • Availability
  • Add to Cart