ADVH49B81F15 AChemBlock

methyl (1R,2S)-2-aminocyclohexanecarboxylate;hydrochloride

  • Cas Number : 835926-44-8
  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1r-2s-2-aminocyclohexanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73914
Availability details for
Quantity
  • Availability
  • Add to Cart