ADVH49B81F17 AChemBlock

(4-Methoxy-3,5-dimethylphenyl)methanol

  • Cas Number : 83037-98-3
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-3-5-dimethylphenyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70445
Availability details for
Quantity
  • Availability
  • Add to Cart