ADVH49B81F19 AChemBlock

5-Ethynyl-2-pyridinamine

  • Cas Number : 82454-61-3
  • Molecular Weight : 118.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-ethynyl-2-pyridinamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86209
Availability details for
Quantity
  • Availability
  • Add to Cart