ADVH49B81F1C AChemBlock

6-[(tert-butoxy)carbonyl]-1H,4H,5H,6H,7H-pyrazolo[3,4-c]pyridine-3-carboxylic acid

  • Cas Number : 821785-76-6
  • Molecular Weight : 267.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-tert-butoxy-carbonyl-1h-4h-5h-6h-7h-pyrazolo-3-4-c-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73957
Availability details for
Quantity
  • Availability
  • Add to Cart