ADVH49B81F1D AChemBlock

3-chloro-6-methoxy-2-methyl-benzaldehyde

  • Cas Number : 82128-14-1
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-6-methoxy-2-methyl-benzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73958
Availability details for
Quantity
  • Availability
  • Add to Cart