ADVH49B81F1F AChemBlock

8-methylquinoline-4-carboxylic acid

  • Cas Number : 816448-09-6
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-methylquinoline-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73971
Availability details for
Quantity
  • Availability
  • Add to Cart