ADVH49B81F26 AChemBlock

3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid

  • Cas Number : 80194-68-9
  • Molecular Weight : 225.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-5-trifluoromethyl-pyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71057
Availability details for
Quantity
  • Availability
  • Add to Cart