ADVH49B81F2A AChemBlock

4,6-dichloro-5-methylsulfonyl-pyrimidin-2-amine

  • Cas Number : 78744-34-0
  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-5-methylsulfonyl-pyrimidin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74068
Availability details for
Quantity
  • Availability
  • Add to Cart