ADVH49B81F2B AChemBlock

ethyl 2-(3-bromophenyl)thiazole-4-carboxylate

  • Cas Number : 786654-97-5
  • Molecular Weight : 312.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-3-bromophenyl-thiazole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71064
Availability details for
Quantity
  • Availability
  • Add to Cart