ADVH49B81F2D AChemBlock

RGB-286638

  • Cas Number : 784210-88-4
  • Molecular Weight : 545.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/rgb-286638.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72356
Availability details for
Quantity
  • Availability
  • Add to Cart