ADVH49B81F36 AChemBlock

(E)-3-cyclopentyl-2-methylacrylic acid

  • Cas Number : 773112-99-5
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/e-3-cyclopentyl-2-methylacrylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71071
Availability details for
Quantity
  • Availability
  • Add to Cart