ADVH49B81F38 AChemBlock

7-(hydroxymethyl)-2,3-dihydro-1H-isoindol-1-one

  • Cas Number : 771-07-3
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-hydroxymethyl-2-3-dihydro-1h-isoindol-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74150
Availability details for
Quantity
  • Availability
  • Add to Cart