ADVH49B81F3A AChemBlock

4-Chloro-2-fluorophenylboronic acid pinacol ester

  • Cas Number : 765917-27-9
  • Molecular Weight : 274.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-2-fluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70615
Availability details for
Quantity
  • Availability
  • Add to Cart