ADVH49B81F41 AChemBlock

3-bromo-5-chloropyridine-2-carbonitrile

  • Cas Number : 760207-83-8
  • Molecular Weight : 217.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-chloropyridine-2-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71086
Availability details for
Quantity
  • Availability
  • Add to Cart