ADVH49B81F43 AChemBlock

2-(3,5-Dibromophenyl)ethanol

  • Cas Number : 75894-93-8
  • Molecular Weight : 279.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-5-dibromophenyl-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70308
Availability details for
Quantity
  • Availability
  • Add to Cart