ADVH49B81F4B AChemBlock

4-methyloct-2-yn-4-ol

  • Cas Number : 74514-59-3
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methyloct-2-yn-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71098
Availability details for
Quantity
  • Availability
  • Add to Cart