ADVH49B81F4C AChemBlock

4-(4-chlorophenoxy)-2-(trifluoromethyl)benzenamine

  • Cas Number : 74159-77-6
  • Molecular Weight : 287.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-chlorophenoxy-2-trifluoromethyl-benzenamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71100
Availability details for
Quantity
  • Availability
  • Add to Cart