ADVH49B81F4D AChemBlock

3-chloro-2,5-diisopropylpyrazine

  • Cas Number : 74152-17-3
  • Molecular Weight : 198.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-2-5-diisopropylpyrazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71101
Availability details for
Quantity
  • Availability
  • Add to Cart