ADVH49B81F4F AChemBlock

3,4'-dimethylbiphenyl

  • Cas Number : 7383-90-6
  • Molecular Weight : 182.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-dimethylbiphenyl.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71104
Availability details for
Quantity
  • Availability
  • Add to Cart