ADVH49B81F51 AChemBlock

(R)-5-(4-amino-3-methyl-phenyl)-8-methyl-7-(5-methyl-1,3,4-thiadiazol-2-yl)-8,9-dihydro-7H-1,3-dioxolo[4,5-h][2,3]-benzodiazepine

  • Cas Number : 732278-52-3
  • Molecular Weight : 407.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/r-5-4-amino-3-methyl-phenyl-8-methyl-7-5-methyl-1-3-4-thiadiazol-2-yl-8-9-dihydro-7h-1-3-dioxolo-4-5-h-2-3-benzodiazepine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86589
Availability details for
Quantity
  • Availability
  • Add to Cart